4-Nitrobenzhydrazide, 98+%
4-Nitrobenzhydrazide, 98+%
4-Nitrobenzhydrazide, 98+%
4-Nitrobenzhydrazide, 98+%
Thermo Scientific Chemicals
4-Nitrobenzhydrazide, 98+%
CAS: 636-97-5 | C7H7N3O3 | 181.151 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18016.0910 g
Catalog number ALFA18016.09
Price (MYR)
420.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
420.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitrobenzhydrazide
CAS636-97-5
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c