Di-n-octyl sulfide, 97%
Di-n-octyl sulfide, 97%
Di-n-octyl sulfide, 97%
Di-n-octyl sulfide, 97%
Thermo Scientific Chemicals
Di-n-octyl sulfide, 97%
CAS: 2690-08-6 | C16H34S | 258.51 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17955.1425 g
Catalog number ALFA17955.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-octyl sulfide
CAS2690-08-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a pharmaceutical intermediate.
Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)
Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. W. V. Steele.; R. D. Chirico.; S. E. Knipmeyer.; A. Nguyen.; and N. K. Smith. Thermodynamic Properties and Ideal-Gas Enthalpies of Formation for Dicyclohexyl Sulfide, Diethylenetriamine, Di-n-octyl Sulfide, Dimethyl Carbonate, Piperazine, Hexachloroprop-1-ene, Tetrakis(dimethylamino)ethylene, N,N‘-Bis-(2-hydroxyethyl)ethylenediamine, and 1,2,4-Triazolo[1,5-a]pyrimidine.J. Chem. Eng. Data.,1997,42(6), 1037-1052.
  2. Hirokazu Narita.; Mikiya Tanaka.; Kazuko Morisaku. Palladium extraction with N,N,N',N'-tetra-n-octyl-thiodiglycolamide.Miner. Eng.,2008,21(6), 483-488.