Tetramethylphosphonium bromide, 97%
Tetramethylphosphonium bromide, 97%
Tetramethylphosphonium bromide, 97%
Tetramethylphosphonium bromide, 97%
Thermo Scientific Chemicals
Tetramethylphosphonium bromide, 97%
CAS: 4519-28-2 | C4H12BrP | 171.02 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17907.065 g
Catalog number ALFA17907.06
Price (MYR)
1,628.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,628.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetramethylphosphonium bromide
CAS4519-28-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as a pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Staffan Andersson and Susan Jagner. Coordination of Copper (∣) in Bis (tetramethylphosphonium) Tribromocuprate (∣) and Bis (tetraethylphosphonium) Di-u-bromo-dibromodicuprate (∣); Comparison od Anionic Configurations in Bromocuprates(∣) Crystallizing with Symmetrical Tetraalkylammonium and Related Cations. Acta chemica Scandinavica. Series A.,1987,41 230-236.
  2. Adel Rafai Far.; Alexander Shivanyuk.; and Julius Rebek, Jr. Water-Stabilized Cavitands.J. Am. Chem. Soc.,2002,124(12), 2854-2855.