Tin(II) oxide, 99%
Tin(II) oxide, 99%
Tin(II) oxide, 99%
Thermo Scientific Chemicals
Tin(II) oxide, 99%
CAS: 21651-19-4 | OSn | 134.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA17889.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTin(II) oxide
Melting Point1080°C (decomposition)
CAS21651-19-4
Health Hazard 1H335-H373
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
View more
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
Solubility
Insoluble in water and alcohol.
Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.
RUO – Research Use Only
General References:
  1. Yamaguchi, H.; Nakanishi, S.; Iba, H.; Itoh, T. Amorphous polymeric anode materials from poly(acrylic acid) and tin(II) oxide for lithium ion batteries. J. Power Sources 2015, 275, 1-5.
  2. Kaizra, S.; Louafi, Y.; Bellal, B.; Trari, M.; Rekhil, G. Electrochemical growth of tin(II) oxide films: Application in photocatalytic degradation of methylene blue. Mater. Sci. Semicond. Process. 2015, 30, 554-560.