Thiodiglycolic acid, 98%
Thiodiglycolic acid, 98%
Thiodiglycolic acid, 98%
Thiodiglycolic acid, 98%
Thermo Scientific Chemicals
Thiodiglycolic acid, 98%
CAS: 123-93-3 | C4H6O4S | 150.148 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17880.22100 g
Catalog number ALFA17880.22
Price (MYR)
540.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
540.00
EA
Add to cart
Specifications
Chemical Name or MaterialThiodiglycolic acid
CAS123-93-3
Melting Point127°C to 131°C
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.
Solubility
Solubility in water is almost transparent. (water, 4.00E+05 mg/L @ 25°C (exp); 1e+006 mg/L @ 25°C (est).
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. L W Wormhoudt.; J N Commandeur.; J H Ploemen.; R S Abdoelgafoer.; A Makansi.; P J Van Bladeren.; N P Vermeulen. Urinary thiodiacetic acid. A selective biomarker for the cytochrome P450-catalyzed oxidation of 1,2-dibromoethane in the rat.Drug Metab Dispos.,1997,25(4), 508-515.
  2. U Hofmann.; M Eichelbaum.; S Seefried.; C O Meese. Identification of thiodiglycolic acid, thiodiglycolic acid sulfoxide, and (3-carboxymethylthio)lactic acid as major human biotransformation products of S-carboxymethyl-L-cysteine.Drug Metab Dispos.,1991,19(1), 222-226.