Gibberellic acid, 90+%
Gibberellic acid, 90+%
Gibberellic acid, 90+%
Gibberellic acid, 90+%
Thermo Scientific Chemicals
Gibberellic acid, 90+%
CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17843.031 g
Catalog number ALFA17843.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGibberellic acid
CAS77-06-5
Health Hazard 1H319
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.
Solubility
Soluble in water 20°C 5g/L. Soluble in Ethanol: 200 mg/4ml.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Claus Schwechheimer. Understanding gibberellic acid signaling--are we there yet?Curr Opin Plant Biol.,2008,11(1), 9-15.
  2. Sven Eriksson.; Henrik Böhlenius.; Thomas Moritz.; Ove Nilsson. GA4 is the active gibberellin in the regulation of LEAFY transcription and Arabidopsis floral initiation.Plant Cell.;2006,18(9), 2172-2181.
  3. Shinjiro Yamaguchi. Gibberellin metabolism and its regulation.Annu Rev Plant Biol.;2008,59225-251.
  4. Plant growth regulator. For an overview of the chemistry of gibberelins, see: Chem. Rev., 92, 573 (1992).