N,N-Diethyl-p-phenylenediamine sulfate, 97%
N,N-Diethyl-p-phenylenediamine sulfate, 97%
N,N-Diethyl-p-phenylenediamine sulfate, 97%
N,N-Diethyl-p-phenylenediamine sulfate, 97%
Thermo Scientific Chemicals
N,N-Diethyl-p-phenylenediamine sulfate, 97%
CAS: 6283-63-2 | C10H18N2O4S | 262.324 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17832.1850 g
Catalog number ALFA17832.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N-Diethyl-p-phenylenediamine sulfate
CAS6283-63-2
Health Hazard 1H302+H312
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
View more
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.
Solubility
Soluble in water and alcohol.
Notes
Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Pachamuthu, M. P.; Karthikeyan, S.; Sekaran, G.; Maheswari, R.; Ramanathan, A. Fenton-Type Oxidative Degradation of N,N-Diethyl-p-phenylenediamine by a Mesoporous Wormhole Structured FeTUD-1 Catalyst. Clean: Soil, Air, Water 2015, 43 (3), 375-381.
  2. Melnikova, L.; Petrenko, V. I.; Avdeev, M. V.; Ivankov, O. I.; Bulavin, L. A.; Garamus, V. M.; Almásy, L.; Mitroova, Z.; Kopcansky, P. SANS contrast variation study of magnetoferritin structure at various iron loading. J. Magn. Magn. Mater. 2015, 377, 77-80.
  3. Criado, P.; Fraschini, C.; Salmieri, S.; Becher, D.; Safrany, A.; Lacroix, M. Evaluation of Antioxidant Cellulose Nanocrystals and Applications in Gellan Gum Films. Ind. Biotechnol. 2015, 11 (1), 59-68.