Cyclooctanol, 97%
Cyclooctanol, 97%
Cyclooctanol, 97%
Thermo Scientific Chemicals
Cyclooctanol, 97%
CAS: 696-71-9 | C8H16O | 128.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17825.1425 g
Catalog number ALFA17825.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclooctanol
CAS696-71-9
Health Hazard 1H227
Health Hazard 2GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
Solubility
Miscible with acetone.
Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.
RUO – Research Use Only
General References:
  1. Oxidation of secondary alcohols to ketones with chromic acid in acetone (Jones' reagent) is exemplified for the preparation of cyclooctanone: Org. Synth. Coll., 5, 310 (1973).
  2. Ren, Y.; Cheng, X.; Yang, K.; Zhu, X.; Li, H.; Wang, Y. Raman study of plastic crystal phase of cyclooctanol under high pressure with different compression rate. J. Mol. Struct. 2015, 1012, 6-10.
  3. Huang, Y. B.; Shen, M.; Wang, X.; Shi, P. C.; Li, H.; Cao, R. Hierarchically micro- and mesoporous metal-organic framework-supported alloy nanocrystals as bifunctional catalysts: Toward cooperative catalysis. J. Catal. 2015, 330, 452-457.