Bromothymol Blue
Bromothymol Blue
Bromothymol Blue
Thermo Scientific Chemicals
Bromothymol Blue
CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17746.0910 g
Catalog number ALFA17746.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBromothymol Blue
Melting Point200°C to 202°C
CAS76-59-5
Recommended StorageAmbient temperatures
RTECS NumberSJ7450000
View more
Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.
Solubility
Soluble in ethanol, ether and alkalis. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Acid-base indicator: pH 6.0 - 7.6.
  2. Suwantarat, N.; Watkins, T.; Lee, R.; Carroll, K. C.; Zhang, S. X. False-positive reaction of L-canavanine glycine bromothymol blue medium with Candida famata. J. Clin. Microbiol. 2014, 52 (4), 1308-1309.
  3. El-Nahhal, I. M.; Zourab, S. M.; Kodeh, F. S.; Abdelsalam, F. H. Sol-gel encapsulation of bromothymol blue pH indicator in presence of Gemini 12-2-12 surfactant. J. Sol-Gel Sci. Technol. 2014, 71 (1), 16-23.