2-Methyl-5-nitroaniline, 98+%
2-Methyl-5-nitroaniline, 98+%
2-Methyl-5-nitroaniline, 98+%
2-Methyl-5-nitroaniline, 98+%
Thermo Scientific Chemicals
2-Methyl-5-nitroaniline, 98+%
CAS: 99-55-8 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17726.22100 g
Catalog number ALFA17726.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-5-nitroaniline
CAS99-55-8
Health Hazard 1H301+H311+H331-H351
Health Hazard 2GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
View more
It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.
Solubility
Insoluble in water. (<0.1 g/100 mL at 19°C). Solubility in methanol, almost transparency
Notes
C.I. 37105. Sensitive to moisture and light. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only
General References:
  1. National Toxicology Program. Bioassay of 5-nitro-o-toluidine for possible carcinogenicity (CAS No. 99-55-8).Natl Cancer Inst Carcinog Tech Rep Ser.1978,1071-125.
  2. Claude A Diehl.; Chad T Jafvert.; Karen A Marley.; Richard A Larson. Surfactant-assisted UV-photolysis of nitroarenes.Chemosphere.2002,46(4), 553-560.
  3. Eva Engel.; Francesco Santarelli.; Rudolf Vasold.; Heidi Ulrich.; Tim Maisch.; Burkhard König.; Michael Landthaler.; Neera V Gopee.; Paul C Howard.; Wolfgang Bäumler. Establishment of an extraction method for the recovery of tattoo pigments from human skin using HPLC diode array detector technology.Anal Chem.2006,78(18),6440-6447.