3-(Methacryloyloxy)propyltrimethoxysilane, 97%
3-(Methacryloyloxy)propyltrimethoxysilane, 97%
Thermo Scientific Chemicals
3-(Methacryloyloxy)propyltrimethoxysilane, 97%
CAS: 2530-85-0 | C10H20O5Si | 248.35 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17714.1850 g
Catalog number ALFA17714.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Boiling Point190°C
Chemical Name or Material3-(Methacryloyloxy)propyltrimethoxysilane
CAS2530-85-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
View more
3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
Solubility
Miscible with water.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Gharbharan, D.; Britsch, D.; Soto, G.; Weed, A. K.; Svec, F.; Zajickova, Z. Tuning preparation conditions towards optimized separation performance of thermally polymerized organo-silica monolithic columns in capillary liquid chromatography. J. Chromatogr. A 2015, 1408, 101-107.
  2. Liu, Y.; Chen, R.; Yuan, D.; Liu, Z.; Meng, M.; Wang, Y.; Meng, J. H. X.; Liu, F, Hu, Z. Thermal-responsive ion-imprinted polymer based on magnetic mesoporous silica SBA-15 for selective removal of Sr(II) from aqueous solution. Colloid. Polym. Sci. 2015, 293 (1), 109-123.