L-Histidine monohydrochloride monohydrate, 99%
L-Histidine monohydrochloride monohydrate, 99%
L-Histidine monohydrochloride monohydrate, 99%
L-Histidine monohydrochloride monohydrate, 99%
Thermo Scientific Chemicals
L-Histidine monohydrochloride monohydrate, 99%
L-Histidine monohydrochloride monohydrate, CAS # 5934-29-2, is a histidine precursor through histidine decarboxylase. | CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
Catalog number ALFA17627.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Histidine monohydrochloride monohydrate
Melting Point∼240°C (decomposition)
CAS5934-29-2
Recommended StorageAmbient temperatures
RTECS NumberMS3119000
View more
L-Histidine monohydrochloride monohydrate is used as a precursor through histidine decarboxylase. It is also used to study the cultures of the human T-lymphoblastic leukemia cell line, modulation of apoptosis. It is also used to enhance the biosynthesis of Lovastatin by cultured aspergillus terreus. It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. It is employed as a component to avoid the risk of contaminating bioproducts with adventitious viruses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• L-Histidine monohydrochloride monohydrate is a histidine precursor via histidine decarboxylase.
Application
• As a precursor of histidine, it can be used to enhance the biosyntyhesis of lovastatin by cultured Aspergillus terreus
• It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar
• It can be employed in laboraotry applications as a component to avoid the risk of contaminating bioproducts with adventitious viruses
• It is also used for laboratory study of cultures of the human T-lymphoblastic leukemia cell line, and modulation of apoptosis
RUO – Research Use Only
General References:
  1. Chan-Huot, M.; Wimperis, S.; Gervais, C.; Bodenhausen, G.; Duma, L. Deuterium MAS NMR Studies of Dynamics on Multiple Timescales: Histidine and Oxalic Acid. ChemPhysChem 2015, 16 (1), 204-215.
  2. Revathi, V.; Rajendran, V. Structural, topographical, linear and non linear optical, electrical and mechanical properties of pure and barium @ amino hydrogen chloride monohydrate single crystal by Slow Evaporation Solution Method. Mater. Lett.2015, 161, 75-78.