Sudan II
Sudan II
Sudan II
Thermo Scientific Chemicals
Sudan II
CAS: 3118-97-6 | C18H16N2O | 276.34 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17613.1425 g
Catalog number ALFA17613.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSudan II
Melting Point156°C to 160°C
CAS3118-97-6
Health Hazard 1H341-H351
Health Hazard 2GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.