Triethylamine hydrochloride, 98%
Triethylamine hydrochloride, 98%
Triethylamine hydrochloride, 98%
Triethylamine hydrochloride, 98%
Thermo Scientific Chemicals
Triethylamine hydrochloride, 98%
CAS: 554-68-7 | C6H16ClN | 137.65 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17318.30250 g
Catalog number ALFA17318.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTriethylamine hydrochloride
CAS554-68-7
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
It finds its application as a protein purification reagent. Triethylamine is used as a competing base for the separation of acidic basic and neutral drugs by reverse-phased high-performance liquid chromatography (HPLC). Triethylamine induces visual disturbances (such as foggy vision) in humans, and is also used in industry as a quenching agent in the ozonolysis of alkenes (e.g. (E)-2-Pentene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as a protein purification reagent. Triethylamine is used as a competing base for the separation of acidic basic and neutral drugs by reverse-phased high-performance liquid chromatography (HPLC). Triethylamine induces visual disturbances (such as foggy vision) in humans, and is also used in industry as a quenching agent in the ozonolysis of alkenes (e.g. (E)-2-Pentene.
Solubility
Soluble in water at 20°C 1440 g/L. Soluble in ethanol, chloroform, and alcohol. Insoluble in ether.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only
General References:
  1. James R. Vaughan, et al. The Preparation of Peptides Using Mixed Carbonic—Carboxylic Acid Anhydrides.J. Am. Chem. Soc.,1952,74(3), 676-678.
  2. P. W. Morgan. Synthesis and Properties of Aromatic and Extended Chain Polyamides.Macromolecules.,1977,10 (6), 1381-1390.