1-Decanol, 98+%
1-Decanol, 98+%
1-Decanol, 98+%
Thermo Scientific Chemicals
1-Decanol, 98+%
CAS: 112-30-1 | C10H22O | 158.285 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17288.AP500 mL
Catalog number ALFA17288.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Decanol
CAS112-30-1
Health Hazard 1H227-H319
Health Hazard 2GHS H Statement
H332-H315-H319-H227
Health Hazard 3P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
View more
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only
General References:
  1. Zhang, M.; Zhao, Y.; Liu, Q.; Yang, L.; Fan, G.; Li, F. A La-doped Mg-Al mixed metal oxide supported copper catalyst with enhanced catalytic performance in transfer dehydrogenation of 1-decanol. Dalton Trans. 2016, 45 (3), 1093-1102.
  2. Echaroj, S.; Santikunaporn, M.; Chavadej, S. Micro-kinetic modeling of the catalytic dehydration of 1-decanol over precipitated gamma-Al2O3. Reac. Kinet. Mech. Cat. 2015, 114 (1), 75-91.