Tetramethylthiuram monosulfide, 97%
Tetramethylthiuram monosulfide, 97%
Tetramethylthiuram monosulfide, 97%
Tetramethylthiuram monosulfide, 97%
Tetramethylthiuram monosulfide, 97%
CAS: 97-74-5 | C6H12N2S3 | 208.356 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA17259.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylthiuram monosulfide
CAS97-74-5
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c