2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
Thermo Scientific Chemicals
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
CAS: 3682-35-7 | C18H12N6 | 312.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA17201.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4,6-Tri(2-pyridyl)-1,3,5-triazine
CAS3682-35-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
RUO – Research Use Only
General References:
  1. Reagent for determination of Fe: Anal. Chem., 31, 1862 (1959).
  2. Gaba, M.; Mohan, C. Design, Synthesis and Biological Evaluation of Novel 1,2,5-Substituted Benzimidazole Derivatives as Gastroprotective Anti-inflammatory and Analgesic Agents. Med. Chem. 2015, 5 (2), 058-063.
  3. Zhang, Y. Z.; Zhao, H. H.; Funck, E.; Dunbar, K. R. A Single-Chain Magnet Tape Based on Hexacyanomanganate(III). Angew. Chem. Int. Ed. 2015, 127 (19), 5675-5679.