5-(4-Dimethylaminobenzylidene)rhodanine 98%
5-(4-Dimethylaminobenzylidene)rhodanine 98%
5-(4-Dimethylaminobenzylidene)rhodanine 98%
5-(4-Dimethylaminobenzylidene)rhodanine 98%
Thermo Scientific Chemicals
5-(4-Dimethylaminobenzylidene)rhodanine 98%
CAS: 536-17-4 | C12H12N2OS2 | 264.361 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17152.065 g
Catalog number ALFA17152.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-(4-Dimethylaminobenzylidene)rhodanine 98%
CAS536-17-4
Melting Point275°C to 280°C
Recommended StorageAmbient temperatures
RTECS NumberVI8090000
View more
Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.
Solubility
Insoluble in water, 993.7(mg/L) at 25°C
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
  1. J W Ludlow.; J A Guikema.; R A Consigli. Use of 5-(4-dimethylaminobenzylidene)rhodanine in quantitating silver grains eluted from autoradiograms of biological material.Anal Biochem.1986,154(1), 104-109.
  2. P Emanuele.; Z D Goodman. A simple and rapid stain for copper in liver tissue.Ann Diagn Pathol. 1988,2,(2), 125-126.
  3. Reagent for detection of Ag, Hg, Cu, Au, Pt and Pd ions.