3-Indoxyl sulfate potassium salt, 97%
3-Indoxyl sulfate potassium salt, 97%
3-Indoxyl sulfate potassium salt, 97%
3-Indoxyl sulfate potassium salt, 97%
Thermo Scientific Chemicals
3-Indoxyl sulfate potassium salt, 97%
CAS: 2642-37-7 | C8H6KNO4S | 251.297 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17079.031 g
Catalog number ALFA17079.03
Price (MYR)
1,430.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,430.00
EA
Add to cart
Specifications
Chemical Name or Material3-Indoxyl sulfate potassium salt
CAS2642-37-7
Melting Point165°C (decomposition)
Recommended StorageStore at -20°C
RTECS NumberNM3200000