Quinine hemisulfate monohydrate, 98+%
Quinine hemisulfate monohydrate, 98+%
Quinine hemisulfate monohydrate, 98+%
Quinine hemisulfate monohydrate, 98+%
Thermo Scientific Chemicals
Quinine hemisulfate monohydrate, 98+%
CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALFA17036.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialQuinine hemisulfate monohydrate
CAS6119-70-6
Health Hazard 1H302+H312+H332-H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H301-H334-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
View more
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
Solubility
Soluble in a mixture of chloroform and absolute alcohol (2:1).
Notes
Light sensitive. Incompatible with strong oxidizing agents, alkalis, ammonia, strong bases and iodine.
RUO – Research Use Only
General References:
  1. Krzeszewski, M.; Gryko, D. T. x-Shaped Bis (areno)-1, 4-dihydropyrrolo [3, 2-b] pyrroles Generated by Oxidative Aromatic Coupling. J. Org. Chem. 2015, 80 (5), 2893-2899.
  2. Janiga, A.; Krzeszewski, M.; Gryko, D. T. Diindolo [2, 3-b: 2', 3'-f] pyrrolo [3, 2-b] pyrroles as Electron-Rich, Ladder-Type Fluorophores: Synthesis and Optical Properties. Chem. Asian J. 2015, 10 (1), 212-218.