Sodium hydrogen carbonate, 99%
Sodium hydrogen carbonate, 99%
Sodium hydrogen carbonate, 99%
Sodium hydrogen carbonate, 99%
Thermo Scientific Chemicals
Sodium hydrogen carbonate, 99%
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17005.36500 g
Catalog number ALFA17005.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydrogen carbonate
CAS144-55-8
pH8.1 to 8.4
Recommended StorageAmbient temperatures
RTECS NumberVZ0950000
View more
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.
Solubility
Soluble in water and methanol. Insoluble in ethanol.
Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Yang, X.; Wu, J.; Mao, X.; Jamison, T. F.; Hatton, T. A. Microwave assisted synthesis of cyclic carbonates from olefins with sodium bicarbonates as the C1 source. Chem. Commun. 2014, 50 (24), 3245-3248.
  2. Maurya, M. R.; Kumar, N. Sodium bicarbonate assisted oxidation, by H2O2, of styrene and cyclohexene using polymer grafted dioxidomolybdenum(VI) complex as a catalyst. J. Mol. Catal. A: Chem. 2015, 406, 204-212.