4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
Thermo Scientific Chemicals
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
CAS: 315-30-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16974.065 g
Catalog number ALFA16974.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine
CAS315-30-0
Health Hazard 1H301-H317-H500
Health Hazard 2GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
View more
Xanthine oxidase inhibitor; decreases uric acid production