Dibromochloromethane, 97+%
Dibromochloromethane, 97+%
Dibromochloromethane, 97+%
Dibromochloromethane, 97+%
Thermo Scientific Chemicals
Dibromochloromethane, 97+%
CAS: 124-48-1 | CHBr2Cl | 208.277 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16938.065 g
Catalog number ALFA16938.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDibromochloromethane
CAS124-48-1
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
It affords bromochlorocarbene under phase-transfer conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It affords bromochlorocarbene under phase-transfer conditions.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
RUO – Research Use Only
General References:
  1. M. Fedoryński.; M. Popławska.; K. Nitschke.; W. Kowalski & M. Mâkosza. A Simple Method for Preparation of Dibromochloromethane and Bromodichloromethane.An International Journal for Rapid Communication of Synthetic Organic Chemistry.,1977,7(4), 287-292.
  2. Y. Yokouchi1.; F. Hasebe.; M. Fujiwara.; H. Takashima.; M. Shiotani.; N. Nishi.; Y. Kanaya.; S. Hashimoto.; P. Fraser.; D. Toom-Sauntry.; H. Mukai.; and Y. Nojiri. Correlations and emission ratios among bromoform, dibromochloromethane, and dibromomethane in the atmosphere.Journal of Geophysical Research: Atmospheres .,2005,110(D23).
  3. Source of bromochlorocarbene in the presence of base and Dibenzo-18-crown-6, A13133. Other conditions give mixtures of carbenes: Synthesis, 783 (1977); Tetrahedron, 33, 363 (1977).