2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
Thermo Scientific Chemicals
2,2':5',2″-Terthiophene, 99%
CAS: 1081-34-1 | C12H8S3 | 248.376 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16748.031 g
Catalog number ALFA16748.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2':5',2''-Terthiophene
CAS1081-34-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point89°C to 94°C
View more
2,2?:5?,2??-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Murat Ates; Nuri Eren. Electrochemical copolymerization of carbazole and 2,2':5'-2″ terthiophene: characterization and micro-capacitor application. Iranian Polymer Journal. 2014, 23 581-589.
  2. Mohammad Shamsuddin Ahmed; Haesang Jeong; Jung-Min You; Seungwon Jeon. Synthesis and characterization of an electrochromic copolymer based on 2,2':5',2″-terthiophene and 3,4-ethylenedioxythiophene. Applied Nanoscience. 2012, 2 133-141.