1-Aminoindane, 99%
1-Aminoindane, 99%
1-Aminoindane, 99%
Thermo Scientific Chemicals
1-Aminoindane, 99%
CAS: 34698-41-4 | C9H12N | 134.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA16585.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-1-Aminoindane
CAS34698-41-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c