HEPES sodium salt, 99%
HEPES sodium salt, 99%
HEPES sodium salt, 99%
HEPES sodium salt, 99%
Thermo Scientific Chemicals
HEPES sodium salt, 99%
CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16516.22100 g
Catalog number ALFA16516.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHEPES sodium salt
CAS75277-39-3
Recommended StorageAmbient temperatures; Store under Nitrogen
EINECS Number278-169-7
TSCAYes
View more
HEPES [N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.
Solubility
It is soluble in water.
Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. H Sands .; LJ Gorey-Feret .; AJ Cocuzza .; FW Hobbs. Biodistribution and metabolism of internally 3H-labeled oligonucleotides. I. Comparison of a phosphodiester and a phosphorothioate. Molecular Pharmacology. 1994, 45, (5), 932-943.
  2. DH Lee .; SY Kim .; JI Hong. A fluorescent pyrophosphate sensor with high selectivity over ATP in water. Angewandte Chemie International Edition. 1994, 43, (36), 4777-4780.