3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
CAS: 149022-15-1 | C8H10ClN3S | 215.70 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16317.065 g
Catalog number ALFA16317.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate
CAS149022-15-1
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Sensitive color reagent for the detection and estimation of water-soluble aldehydes: Anal. Chem., 33, 93, 722, 1574 (1961).
  2. Shi, J. Y; Liu, N.; Gu, R. X.; Zhu, L. Q.; Zhang, C.; Wang, Q. G.; Lei, Z. H.; Liu, Y. Y.; Ren, J. Y. Signals induced by exogenous nitric oxide and their role in controlling brown rot disease caused by Monilinia fructicola in postharvest peach fruit. J. Gen. Plant Pathol. 2015, 81 (1), 68-76.
  3. El-Sayed, A. S. A.; Abdel-Azeim, S.; Ibrahim, H. M.; Yassin, M. A.; Abdel-Ghany, S. E.; Esener, S.; Ali, G. S. Biochemical stability and molecular dynamic characterization of Aspergillus fumigatus cystathionine γ-lyase in response to various reaction effectors. Enzyme Microb. Technol. 2015, 81, 31-46.