Hydrocortisone, 98%
Hydrocortisone, 98%
Hydrocortisone, 98%
Thermo Scientific Chemicals
Hydrocortisone, 98%
CAS: 50-23-7 | C21H30O5 | 362.466 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA16292.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrocortisone
CAS50-23-7
Health Hazard 1H360Df-H373
Health Hazard 2GHS H Statement
H361
Health Hazard 3P201-P202-P260-P281-P308+P313-P501c
View more
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.
Solubility
It is soluble in DMSO and ethanol.
Notes
Protect against light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. S Spireas .; S Sadu, R Grover. In vitro release evaluation of hydrocortisone liquisolid tablets. Journal of Pharmaceutical Sciences. 1998, 87, (7), 867-872.
  2. P Dandona.; K Thusu, R Hafeez.; E Abdel-Rahman. Effect of hydrocortisone on oxygen free radical generation by mononuclear cells. Metabolism. 1998, 47, (7), 788-791.