Copper, powder, 99+%
Copper, powder, 99+%
Actual product may vary
Copper, powder, 99+%
Copper, powder, 99+%
Thermo Scientific Chemicals
Copper, powder, 99+%
CAS: 7440-50-8 | Cu
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number ALFA16243.36
Price (MYR)
721.00
EA
Add to cart
Quantity:
500 g
Price (MYR)
721.00
EA
Add to cart
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS7440-50-8
Melting Point1083°C
Boiling Point2595°C
Quantity500 g
Molecular FormulaCu
Percent Purity≥99%
Unit SizeEA