Vitamin A acetate in gelatin, 500,000 I.U./g
Vitamin A acetate in gelatin, 500,000 I.U./g
Vitamin A acetate in gelatin, 500,000 I.U./g
Vitamin A acetate in gelatin, 500,000 I.U./g
Thermo Scientific Chemicals
Vitamin A acetate in gelatin, 500,000 I.U./g
CAS: 127-47-9 | C22H32O2 | 328.496 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16237.1425 g
Catalog number ALFA16237.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialVitamin A acetate
Name Notein gelatin
CAS127-47-9
Health Hazard 1H360D
Health Hazard 2GHS H Statement
H361
Suspected of damaging fertility or the unborn child.