2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
Thermo Scientific Chemicals
2',7'-Dichlorofluorescein
CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16212.065 g
Catalog number ALFA16212.06
Price (MYR)
466.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
466.00
EA
Add to cart
Specifications
Chemical Name or Material2′, 7′-Dichlorofluorescein
CAS76-54-0
Health Hazard 1H315-H319-H335-H500
Health Hazard 2GHS H Statement
H319
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2',7'-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′,7′-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.
Solubility
Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water.
Notes
As a fluorescent indicatorIncompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Liu, X. Y.; Liu, D. Y.; Qi, J.; Cui, Z. G.; Chang, H. X.; He, H. R.; Yang, G. M. A new fluorescent sensor for Cd2+ and its application in living cells imaging. Tetrahedron Lett. 2015, 56 (11), 1322-1327.
  2. Kim, H. Y.; Im, H. G.; Chang, S. K. Colorimetric and fluorogenic signaling of fluoride ions by thiophosphinated dichlorofluorescein. Dyes Pigm. 2015, 112, 170-175.