Copper(II) acetate monohydrate, 98+%
Copper(II) acetate monohydrate, 98+%
Copper(II) acetate monohydrate, 98+%
Copper(II) acetate monohydrate, 98+%
Thermo Scientific Chemicals
Copper(II) acetate monohydrate, 98+%
CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16203.30250 g
Catalog number ALFA16203.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper(II) acetate monohydrate
Melting Point115°C
CAS6046-93-1
Health Hazard 1H302-H314-H335-H373
Health Hazard 2GHS H Statement
H302-H315-H319-H335
View more
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Nagababu, P.; Barui, A. K.; Thulasiram, B.; Devi, C. S.; Satyanarayana, S.; Patra, C. R.; Sreedhar, B. Antiangiogenic Activity of Mononuclear Copper (II) Polypyridyl Complexes for the Treatment of Cancers. J. Med. Chem. 2015, 58 (13), 5226-5241.
  2. Gogoi, K.; Deka, H.; Kumar, V.; Mondal, B. Oxo Transfer from Nitrogen Dioxide to Nitrito Group in a Copper(II) Complex. Inorg. Chem. 2015, 54 (10), 4799-4805.