Barium diphenylamine-4-sulfonate
Barium diphenylamine-4-sulfonate
Barium diphenylamine-4-sulfonate
Barium diphenylamine-4-sulfonate
Thermo Scientific Chemicals
Barium diphenylamine-4-sulfonate
CAS: 6211-24-1 | C24H20BaN2O6S2 | 633.88 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16197.065 g
Catalog number ALFA16197.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBarium diphenylamine-4-sulfonate
CAS6211-24-1
Health Hazard 1H302+H332
Health Hazard 2GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
View more
Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.
Solubility
It is moderate soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with mineral acids, organic acids, strong oxidizing agents.
RUO – Research Use Only
General References:
  1. JJ Antkowiak.; AL Spatorico. Thin-layer chromatography of drugs in medicated feeds. Journal of Chromatography A. 1967, 29, 277-282.
  2. M Qureshi.; SZ Qureshi. A sensitive and selective resin spot test for diphenylamine. Anal. Chem. 1966, 38, (13), 1956-1958.