Zinc oxide, heavy, 98%
Zinc oxide, heavy, 98%
Zinc oxide, heavy, 98%
Zinc oxide, heavy, 98%
Thermo Scientific Chemicals
Zinc oxide, heavy, 98%
CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16188.0B1000 g
Catalog number ALFA16188.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc oxide
Melting Point1975°C
Name Noteheavy
CAS1314-13-2
Recommended StorageAmbient temperatures
View more
Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong alkaline earth metal. Alkali metals.
RUO – Research Use Only
Priyanka Jood,; Rutvik J. Mehta,; Yanliang Zhang,; Theo Borca-Tasciuc,; Shi Xue Dou,; David J. Singhd,; Ganpati Ramanath. Heavy element doping for enhancing thermoelectric properties of nanostructured zinc oxide. RSC Adv.,. 2014, volume (4), 6363-6368.