Sodium tetraborate decahydrate, 99+%
Sodium tetraborate decahydrate, 99+%
Sodium tetraborate decahydrate, 99+%
Sodium tetraborate decahydrate, 99+%
Thermo Scientific Chemicals
Sodium tetraborate decahydrate, 99+%
CAS: 1303-96-4 | B4Na2O7·10H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
10,000 g
Catalog number ALFA16176.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium tetraborate decahydrate
Melting Point75°C
CAS1303-96-4
Health Hazard 1H319-H360FD
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
View more
It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.
Solubility
Soluble in water, glycerol and ethanol. Insoluble in acids
Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Nasir, M. Z. M.; Sofer, Z.; Ambrosi, A.; Pumera, M. A limited anodic and cathodic potential window of MoS2: limitations in electrochemical applications. Nanoscale 2015, 7 (7), 3126-3129.
  2. Hussain, S.; Bharadwaj, S. K.; Chaudhuri, M. K.; Kalita, H. Borax as an Efficient Metal-Free Catalyst for Hetero-Michael Reactions in an Aqueous Medium. Eur. J. org. Chem. 2007, 2007 (2), 374-378.