(+/-)-3-Chloro-1,2-propanediol, 98%
(+/-)-3-Chloro-1,2-propanediol, 98%
(+/-)-3-Chloro-1,2-propanediol, 98%
(+/-)-3-Chloro-1,2-propanediol, 98%
Thermo Scientific Chemicals
(+/-)-3-Chloro-1,2-propanediol, 98%
CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16143.30250 g
Catalog number ALFA16143.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-3-Chloro-1,2-propanediol
CAS96-24-2
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H300-H360-H351-H318-H312-H332-H315-H335
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing cancer.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.