alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
Thermo Scientific Chemicals
alpha-Naphtholbenzein
CAS: 145-50-6 | C27H18O2 | 374.439 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA16125.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialalpha-Naphtholbenzein
CAS145-50-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).
Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.
RUO – Research Use Only
General References:
  1. Acid-base indicator: pH 0.0 - 0.8; 8.2 - 10.0.
  2. Wang, J.; Xu, C.; Song, Q. Rapid and Environmentally Friendly Three-Dimensional-Printed Flow Injection Analysis System for the Determination of the Acid Number in Thermal Conductive Oil. Energy Fuels 2015, 29 (2), 1040-1044.
  3. Özçeşmeci, M. Synthesis, photophysical and photochemical properties of metal-free and zinc(II) phthalocyanines bearing alpha-naphtholbenzein units. J. Organomet. Chem. 2014, 767, 16-21.