PIPES, 98%
PIPES, 98%
PIPES, 98%
Thermo Scientific Chemicals
PIPES, 98%
CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA16090.22
Price (MYR)
673.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
673.00
EA
Add to cart
Specifications
Chemical Name or MaterialPIPES
CAS5625-37-6
Melting Point>300°C
Recommended StorageAmbient temperatures
EINECS Number227-057-6
View more
PIPES is commonly used as a buffering agent in biochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
PIPES is commonly used as a buffering agent in biochemistry.
Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Biological buffer, pKa = 6.8 at 20°: Biochemistry, 5, 467 (1966).
  2. Zeddies, S.; De Cuyper, I. M.; Meer, P. F.; Daal, B. B., Korte, D.; Gutiérrez, L.; Thijssen-Timmer, D. C. Pathogen reduction treatment using riboflavin and ultraviolet light impairs platelet reactivity toward specific agonists in vitro. Transfusion 2014, 54 (9), 2292-2300
  3. Ohta, S.; Sakaguchi, S.; Kobayashi, Y.; Mizuno, N.; Tago, K.; Itoh, H. Agonistic Antibodies Reveal the Function of GPR56 in Human Glioma U87-MG Cells. Biol. Pharm. Bull. 2015, 38 (4), 594-600.