1-Bromotetradecane, 98%
1-Bromotetradecane, 98%
1-Bromotetradecane, 98%
1-Bromotetradecane, 98%
Thermo Scientific Chemicals
1-Bromotetradecane, 98%
CAS: 112-71-0 | C14H29Br | 277.29 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16082.36500 g
Catalog number ALFA16082.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromotetradecane
CAS112-71-0
Melting Point4°C to 6°C
Recommended StorageAmbient temperatures
Density1.018
View more
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Katrin Wedeking; Zhongcheng Mu; Gerald Kehr; Roland Frohlich; Gerhard Erker; Lifeng Chi; Harald Fuchs. Tetradecylferrocene: ordered molecular array of an organometallic amphiphile in the crystal and in a two-dimensional assembled structure on a surface. Langmuir. 2006, 22, (7),3161-3165
  2. Nusrat Jahan, Nawal Paul, Christian J.Petropolis, D.Gerrard Marangoni, T.Bruce Grindley. Synthesis of surfactants based on pentaerythritol. I. Cationic and zwitterionic gemini surfactants. Journal of Organic Chemistry. 2009, 74, (20),7762-7773