Cyclohexane, 99+%
Cyclohexane, 99+%
Cyclohexane, 99+%
Thermo Scientific Chemicals
Cyclohexane, 99+%
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16070.AU1000 mL
Catalog number ALFA16070.AU
Price (MYR)
365.00
EA
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (MYR)
365.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclohexane
Melting Point6.5°C
CAS110-82-7
Health Hazard 1H225-H304-H315-H320-H336
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
View more
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.
Solubility
Miscible with ether, alcohol, acetone and olive oil. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zou, G.; Zhong, W.; Xu, Q.; Xiao, J.; Liu, C.; Li, Y.; Mao, L.; Kirk, S.; Yin, D. Oxidation of cyclohexane to adipic acid catalyzed by Mn-doped titanosilicate with hollow structure. Catal. Commun. 2015, 58, 46-52.
  2. Huang, G.; Mo, L. Q.; Cai, J. L.; Cao, X.; Peng, Y.; Guo, Y. A.; Wei, S. J. Environmentally friendly and efficient catalysis of cyclohexane oxidation by iron meso-tetrakis(pentafluorophenyl) porphyrin immobilized on zinc oxide. Appl. Catal., B 2015, 162, 364-371.