Diethylene glycol monomethyl ether, 98%
Diethylene glycol monomethyl ether, 98%
Diethylene glycol monomethyl ether, 98%
Diethylene glycol monomethyl ether, 98%
Thermo Scientific Chemicals
Diethylene glycol monomethyl ether, 98%
CAS: 111-77-3 | C5H12O3 | 120.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16063.AP500 mL
Catalog number ALFA16063.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylene glycol monomethyl ether
Melting Point-70°C
Name Notestabilized with 50 to 150mg/kg BHT
CAS111-77-3
Health Hazard 1H227-H311-H319-H361d
View more
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
RUO – Research Use Only
General References:
  1. Toshihiro Kawamoto.; Koji Matsuno.; Fujio Kayama.; Manabu Hirai.; Keiichi Arashidani.; Yoshikawa Masahiro.; Yasushi Kodama. Effect of ethylene glycol monomethyl ether and diethylene glycol monomethyl ether on hepatic metabolizing enzymes. Toxicology. 1990, 62 (3), 265-274.
  2. Amalendu Pal.; Yoginder P. Singh. Excess Molar Volumes and Viscosities for Glycol Ether-Water Solutions at the Temperature 308.15 K: Ethylene Glycol Monomethyl, Diethylene Glycol Monomethyl, and Triethylene Glycol Monomethyl Ethers. J. Chem. Eng. Data. 1996, 41 (3), 425-427.