Methylcyclohexane, 99%
Methylcyclohexane, 99%
Methylcyclohexane, 99%
Methylcyclohexane, 99%
Thermo Scientific Chemicals
Methylcyclohexane, 99%
CAS: 108-87-2 | C7H14 | 98.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16057.AP500 mL
Catalog number ALFA16057.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMethylcyclohexane
CAS108-87-2
Health Hazard 1H225-H304-H315-H319-H335-H336
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
View more
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. For use as a solvent in the preparation of unsolvated Grignard reagents from an alkyl halide and Mg powder: Org. Synth. Coll., 5, 1141 (1973).
  2. Narayanaswamy, K.; Pitsch, H.; Pepiot, P. A chemical mechanism for low to high temperature oxidation of methylcyclohexane as a component of transportation fuel surrogates. Combust. Flame 2015, 162 (4), 1193-1213.
  3. Ngoye, F.; Lakiss, L.; Qin, Z.; Laforge, S.; Canaff, C.; Tarighi, M.; Valtchev, V.; Thomas, K.; Vicente, A.; Gilson, J. Mitigating coking during methylcyclohexane transformation on HZSM-5 zeolites with additional porosity. J. Catal. 2014, 320, 118-126.