2-Chloro-5-(trifluoromethyl)aniline, 97%
2-Chloro-5-(trifluoromethyl)aniline, 97%
2-Chloro-5-(trifluoromethyl)aniline, 97%
2-Chloro-5-(trifluoromethyl)aniline, 97%
Thermo Scientific Chemicals
2-Chloro-5-(trifluoromethyl)aniline, 97%
CAS: 121-50-6 | C7H5ClF3N | 195.569 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16004.1425 g
Catalog number ALFA16004.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Chloro-5-(trifluoromethyl)aniline
CAS121-50-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.