4,7-Dichloroquinoline, 98%
4,7-Dichloroquinoline, 98%
4,7-Dichloroquinoline, 98%
4,7-Dichloroquinoline, 98%
Thermo Scientific Chemicals
4,7-Dichloroquinoline, 98%
CAS: 86-98-6 | C9H5Cl2N | 198.046 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15985.36500 g
Catalog number ALFA15985.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material4,7-Dichloroquinoline
CAS86-98-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Alexander R. Surrey.; Royal A. Cutler. The Role of Phenol in the Reaction of 4,7-Dichloroquinoline with Novol Diamine. J. Am. Chem. Soc. 1951, 73 (6), 2623-2626.
  2. Robert C. Elderfield.; Walter J. Gensler.; Oskar Birstein.; Frank J. Kreysa.; John T. Maynard.; Jean Galbreath. Synthesis of Certain Simple 4-Aminoquinoline Derivatives. J. Am. Chem. Soc. 1946, 68 (7), 1250-1251.