1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
Thermo Scientific Chemicals
1,8-Diaminonaphthalene, 97%
CAS: 479-27-6 | C10H10N2 | 158.204 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15966.1850 g
Catalog number ALFA15966.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1,8-Diaminonaphthalene
CAS479-27-6
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H301-H351-H317
Toxic if swallowed.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
View more
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Xin-Gui Li.; Mei-Rong Huang.; Sheng-Xian Li. Facile synthesis of poly(1,8-diaminonaphthalene) microparticles with a very high silver-ion adsorbability by a chemical oxidative polymerization. Acta Materialia. 2004, 52 (18), 5363-5374 .
  2. B.J. Palys.; J. Bukowska.; K. Jackowska. SERS of 1,8-diaminonaphthalene on gold, silver and copper electrodes Polymerisation and complexes formed with the electrode material. Journal of Electroanalytical Chemistry. 1997, 428 (1-2), 19-24.