Thiomorpholine, 98%
Thiomorpholine, 98%
Thiomorpholine, 98%
Thermo Scientific Chemicals
Thiomorpholine, 98%
CAS: 123-90-0 | C4H9NS | 103.183 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15958.065 g
Catalog number ALFA15958.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThiomorpholine
CAS123-90-0
Health Hazard 1H227-H314
Health Hazard 2GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.
Solubility
Miscible with waterMiscible with water and organic solvents.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.
RUO – Research Use Only
General References:
  1. Tooulia, K. K.; Theodosis-Nobelos, P.; Rekka, E. A. Thiomorpholine Derivatives with Hypolipidemic and Antioxidant Activity. Arch. Pharm. 2015, 348 (9), 629-634.
  2. Kacan, M.; Turkyilmaz, M.; Karabulut, F.; Altun, O.; Baran, Y. Complexation, thermal and catalytic studies of N-substituted piperazine, morpholine and thiomorpholine with some metal ions. Spectrochim. Acta, Part A 2014, 118, 572-577.