5-Methylbenzimidazole, 98%
5-Methylbenzimidazole, 98%
5-Methylbenzimidazole, 98%
5-Methylbenzimidazole, 98%
Thermo Scientific Chemicals
5-Methylbenzimidazole, 98%
CAS: 614-97-1 | C8H8N2 | 132.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA15957.22
Price (MYR)
2,335.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,335.00
EA
Add to cart
Specifications
Chemical Name or Material5-Methylbenzimidazole
CAS614-97-1
Health Hazard 1H302
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.
Solubility
Soluble in chloroform (25mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. M Tahir Güllüoglu.; Mustafa Ozduran.; Mustafa Kurt.; S Kalaichelvan.; N Sundaraganesan.; M. Tahir Güllüoglu.; Mustafa Özduran.; Mustafa Kurt.; S. Kalaichelvan.; N. Sundaraganesan. Molecular structure and vibrational spectra of 2- and 5-methylbenzimidazole molecules by density functional theory. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2010, 76 (2), 107-114 .
  2. WRIGHT JB. The chemistry of the benzimidazoles. Chem Rev. 1951, 48 (3), 397-541.