Choline chloride, 98+%
Choline chloride, 98+%
Choline chloride, 98+%
Choline chloride, 98+%
Thermo Scientific Chemicals
Choline chloride, 98+%
CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number ALFA15828.36
Price (MYR)
490.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
490.00
EA
Add to cart
Specifications
Chemical Name or MaterialCholine Chloride
CAS67-48-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
View more
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
RUO – Research Use Only
General References:
  1. Sun, S.; Niu, Y.; Xu, Q.; Sun, Z.; Wei, X. Efficient SO2 Absorptions by Four Kinds of Deep Eutectic Solvents Based on Choline Chloride. Ind. Eng. Chem. Res. 2015, 54 (33), 8019-8024.
  2. Das, A.; Biswas, R. Dynamic Solvent Control of a Reaction in Ionic Deep Eutectic Solvents: Time-Resolved Fluorescence Measurements of Reactive and Nonreactive Dynamics in (Choline Chloride + Urea) Melts. J. Phys. Chem. B 2015, 119 (31), 10102-10113.