meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
Thermo Scientific Chemicals
meso-Erythritol, 99%
CAS: 149-32-6 | C4H10O4 | 122.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15813.1425 g
Catalog number ALFA15813.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialmeso-Erythritol
CAS149-32-6
Melting Point119°C to 124°C
Recommended StorageAmbient temperatures
RTECS NumberKF2000000
View more
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.
Solubility
Soluble in water (0.1 g/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at -20°C.
RUO – Research Use Only
General References:
  1. Emmanuel Jean Teinkela Mbosso.; René Wintjens.; Bruno Ndjakou Lenta.; Silvère Ngouela.; Michel Rohmer.; Etienne Tsamo. Chemical constituents from Glyphaea brevis and Monodora myristica: chemotaxonomic significance. Chemistry & Biodiversity. 2013, 10 (2), 224-232.
  2. Freek Spitaels.; Anneleen Wieme.; Tom Balzarini.; Ilse Cleenwerck.; Anita Van Landschoot.; Luc De Vuyst.; Peter Vandamme. Gluconobacter cerevisiae sp. nov., isolated from the brewery environment. International Journal of Systematic and Evolutionary Microbiology. 2014, 64 (4), 1134-1141.