2,2'-Bipyridine, 99+%
2,2'-Bipyridine, 99+%
2,2'-Bipyridine, 99+%
2,2'-Bipyridine, 99+%
Thermo Scientific Chemicals
2,2'-Bipyridine, 99+%
CAS: 366-18-7 | C10H8N2 | 156.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA15782.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2'-Bipyridine
CAS366-18-7
Health Hazard 1H301-H312-H500
Health Hazard 2GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.
Health Hazard 3P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c
View more
Reagent for iron determinations.2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for iron determinations.2,2′-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.
Solubility
Soluble in organic solvents and slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition.
RUO – Research Use Only
General References:
  1. Reagent for iron. Chelating ligand.
  2. For use as an alternative to phosphines in Heck type reactions, see: Synlett, 871 (1992).
  3. Complex with Chromium(VI) oxide, 12522, has been used in a mild selective oxidation of alcohols to aldehydes and ketones: Acta Chem. Scand., 25, 1125 (1971).
  4. Valvekens, P.; Bloch, E. D.; Long, J. R.; Ameloot, R.; De Vos, D. E. Counteranion effects on the catalytic activity of copper salts immobilized on the 2,2'-bipyridine-functionalized metal-organic framework MOF-253. Catal. Today 2015, 246, 55-59.
  5. Kageshima, Y.; Kumagai, H.; Minegishi, T.; Kubota, J.; Domen, K. A Photoelectrochemical Solar Cell Consisting of a Cadmium Sulfide Photoanode and a Ruthenium-2,2'-Bipyridine Redox Shuttle in a Non-aqueous Electrolyte. Angew. Chem. 2015, 127 (27), 7988-7992.